Learn More
3M™ Reusable Earplugs
The Earplugs 1200 Series are reusable and designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up to 26 dB.
Brand: 3M™ 7100288286
2391.33 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Get comfortable and durable hearing protection with 3M™ Earplugs 1200 Series.
- These reusable earplugs reduce noise levels up to 26 dB and are fully tested and approved against EN 352-2:2020.
- They are made from a soft durable Thermoplastic Elastomer (TPE).
- The specially designed and triple flange shape is tapered to fit most ear canal sizes, offering a comfortable secure fit.
- The finger grip design keeps insertion and removal simple, convenient and hygienic.
- Packed in a reusable recyclable cardboard pillow pack, helping keep your earplugs hygienic and together, preventing loss.
- A reusable Polycon plastic box is available separately.
- Allowing you the option of a more robust storage solution, but only buying what you need and potentially reducing plastic wastage.
- 3M™ Earplugs 1261 are available as probed versions for use with the 3M™ E-A-RFit™ validation system.
Specifications
Regular | |
26 dB | |
Flanged | |
No | |
No |
1261 | |
Uncorded | |
Reusable | |
No |
Product Suggestions
Customers who viewed this item also viewed.
CE_17883256_EN.pdf
File size: 820.3 kB
Product Content Correction
Product Title
3M™ Reusable Earplugs > Uncorded
Spot an opportunity for improvement?Share a Content Correction