Learn More
Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths
Allows two shelf depths to accommodates shallow vessels. Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths can be used with a variety of other Grant Heating Circulators and Heated Circulating Baths.
1335.00 DKK - 2400.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
11456407
|
Grant Instruments™
RS18H |
Half the area of SBB Aqua 18 Plus or Sub Aqua 18 Pro |
1565.00 DKK
Each |
|
||||||
11466407
|
Grant Instruments™
RS28H |
Half the area of SBB Aqua 26 Plus or Sub Aqua 26 Pro |
1865.00 DKK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
11446407
|
Grant Instruments™
RS14H |
Half the area of SBB Aqua 12 Plus, Sub Aqua 12 Pro, or Sub Aqua Pro Dual |
1335.00 DKK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
||||||
11476407
|
Grant Instruments™
RS36H |
Half the area of SBB Aqua 34 Plus or Sub Aqua 34 Pro |
2400.00 DKK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
Description
- Choose from a variety of sizes to fit your tank.
- For use with either Stainless Steel and Plastic tank models
Specifications
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Grant Instruments™ Reversible Raised Shelf for Unstirred Water Baths