missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals RGL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00023179-D01P
This item is not returnable.
View return policy
Description
RGL1 Polyclonal antibody specifically detects RGL1 in Human samples. It is validated for Western BlotSpecifications
RGL1 | |
Polyclonal | |
Western Blot 1:500 | |
NP_055964.3 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS (pH 7.4) | |
KIAA0959, ral guanine nucleotide dissociation stimulator-like 1, RGLRalGDS-like 1 | |
RGL1 (NP_055964.3, 1 a.a. - 803 a.a.) full-length human protein. MEVKPVGEPTQEVSKFKLSTKVESTGHWLVEDHVRIWEVLKTEESSIQDWGEEVEEGAVYHVTLKRVQIQQAANKGARWLGVEGDQLPPGHTVSQYETCKIRTIKAGTLEKLVENLLTAFGDNDFTYISIFLSTYRGFASTKEVLELLLDRYGNLTSPNCEEDGSQSSSESKMVIRNAIASILRAWLDQCAEDFREPPHFPCLQKLLDYLTRMMPGSDPERRAQNLLEQFQKQEVETDNGLPNTISFSLEEEEELEGGESAEFTCFSEDLVAEQLTYMDAQLFKKVVPHHCLGCIWSRRDKKENKHLAPTIRATISQFNTLTKCVVSTILGGKELKTQQRAKIIEKWINIAHECRLLKNFSSLRAIVSALQSNSIYRLKKTWAAVPRDRMLMFEELSDIFSDHNNHLTSRELLMKEGTSKFANLDSSVKENQKRTQRRLQLQKDMGVMQGTVPYLGTFLTDLTMLDTALQDYIEGGLINFEKRRREFEVIAQIKLLQSACNSYCMTPDQKFIQWFQRQQLLTEEESYALSCEIEAAADASTTSPKPRKSMVKRLSLLFLGSDMITSPTPTKEQPKSTASGSSGESMDSVSVSSCESNHSEAEEGSITPMDTPDEPQKKLSESSSSCSSIHSMDTNSSGMSSLINPLSSPPSCNNNPKIHKRSVSVTSITSTVLPPVYNQQNEDTCIIRISVEDNNGNMYKSIMLTSQDKTPAVIQRAMLKHNLDSDPAEEYELVQVISEDKELVIPDSANVFYAMNSQVNFDFILRKKNSMEEQVKLRSRTSLTLPRTAKRGCWSNRHSKITL | |
0.1 mg | |
Signal Transduction | |
23179 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |