Learn More
Thermo Scientific™ Rheodyne™ 7010 Sample Injector
Ensure accurate and precise high-volume HPLC injections with the single-mode Thermo Scientific™ Rheodyne™ 7010 Sample Injector.
Brand: Thermo Scientific™ 7010
11507.80 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Ensure accurate and precise high-volume HPLC injections of 5μL to 20mL samples with the Thermo Scientific™ Rheodyne™ 7010 Sample Injector. This single-mode injector uses the complete filling method to load the sample loop.
Specifications
Single Mode | |
Sample Injector | |
Stainless Steel |
5 ÎĽL to 5 mL | |
6 Ports |