missing translation for 'onlineSavingsMsg'
Learn More

Ro 67-4853, Tocris Bioscience™

Product Code. 18064542
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Positive allosteric modulator of group I mGlu receptors

Positive allosteric modulator of mGlu group I receptors (pEC50 value is 7.16 for the rat mGlu1a receptor). Exhibits activity at all group I receptors, including human and rat mGlu1 and rat mGlu5. Enhances the effects of (S)-DHPG in CA3 neurons (EC50 = 95 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302841-89-0
Molecular Formula C19H19NO4
Molecular Weight (g/mol) 325.364
InChI Key RQBUXEUMZZQUFY-UHFFFAOYSA-N
Synonym butyl n-9h-xanthene-9-carbonyl carbamate, butyl 9h-xanthene-9-carbonylcarbamate, 9h-xanthene-9-carbonyl-carbamic acid butyl ester, carbamic acid, 9h-xanthen-9-ylcarbonyl-, butyl ester, d0e6gg, ro hplc, 9h-xanthen-9-ylcarbonyl-carbamic acid butyl ester
PubChem CID 9949202
IUPAC Name butyl N-(9H-xanthene-9-carbonyl)carbamate
SMILES CCCCOC(=O)NC(=O)C1C2=CC=CC=C2OC3=CC=CC=C13

Specifications

Quantity 10 mg
Formula Weight 325.36
Percent Purity >99%
Chemical Name or Material Ro 67-4853
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.