missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ROS1 Recombinant Human Protein
ROS1 is a tyrosine protein kinase.
13290.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_002935 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 79.6kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15408553
|
Invitrogen™
PR7292A |
100μg |
13290.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10211163
|
Invitrogen™
PV4120 |
1mg |
57500.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROS1 functions as a growth or differentiation factor receptor.
Specifications
NP_002935 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
SEV Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
79.6kDa | |
RUO - research use only | |
ROS1, ROS, MCF4 | |
Human | |
GST Tag | |
Frozen |