Learn More
Eppendorf™ Rotor Lids
Designed for use with Eppendorf Rotors
Brand: Eppendorf™ 5409703005
1950.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement rotor lids
- Aerosol-tight and aluminum lids available
- Available in a variety of sizes
Specifications
Rotor Lid | |
Aluminum | |
FA-45-24-11 | |
Aerosol-tight |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Product Title
Eppendorf™ Rotor Lids > For FA-45-24-11; Aerosol-Tight
Spot an opportunity for improvement?Share a Content Correction