Learn More
Cowie Technology™ Round-Bottom Flask, PTFE
COWIE TECHNOLOGY™ Round-Bottom Flask, PTFE has a smooth internal finish and is usable at high temperatures.
Brand:  Cowie Technology™ 040.050.24
 View more versions of this product
Product Code. 10784903
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Round Bottom Flasks have a smooth internal finish.
  • PTFE used in the manufacture of Cowie™ PTFE Laboratory Products is pure, virgin material that conforms to FDA and USP Class VI requirements.
  • Inert and usable to 280°C.
  • Tapers match standard ground glass joints.
  • Without sealing rings.
Specifications
Specifications
52 mm
PTFE
Up to 280°C
50 mL
24/29