missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Duran™ Round Bottom Narrow Necked Flask
These flasks are ideal for boiling and heating applications due to uniform wall thickness distribution.
Brand: Duran™ 217921579
This item is not returnable.
View return policy
Description
- Promotes uniform heating
- Manufactured from clear borosilicate glass 3.3
- Outstanding chemical resistance and near inert behavior
- High temperature and thermal shock resistance
Specifications
Clear | |
Borosilicate Glass (3.3) | |
Narrow | |
Round |
50,000 mL | |
Boiling, heating | |
Round bottom flask |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Duran™ Round Bottom Narrow Necked Flask > Capacity: 50000 mL
Spot an opportunity for improvement?Share a Content Correction