Learn More
Bochem™ Round Wire Basket
Bochem™ Round Wire Basket is constructed of high quality Stainless Steel and includes a handle.
Brand:  Bochem™ 10052
Product Code. 10326291
  • 533.00 DKK / Each
Only null left
Estimated Shipment: 11-09-2025
Description
  • Round
  • Special high quality processing
  • Stainless Steel
Specifications
Specifications
18/10 E-Poli
120 mm
150 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bochem™ Round Wire Basket > Length: 120mm