missing translation for 'onlineSavingsMsg'
Learn More

RPL29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen RPL29
Concentration 0.14 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16835144
View Documents
Proteintech
15799-1-AP-20UL
20 μL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
16825144
View Documents
Proteintech
15799-1-AP-150UL
150 μL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Dual specificity protein phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the MAPK superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of this family of dual specificity phosphatases show distinct substrate specificities for MAPKs, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product binds to and inactivates p38 and SAPK/JNK, but not MAPK/ERK. Its subcellular localization is unique; it is evenly distributed in both the cytoplasm and the nucleus. This gene is widely expressed in various tissues and organs, and its expression is elevated by stress stimuli. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications

Specifications

RPL29
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
P25886, P47914, P47915
19944, 29283, 6159
RPL29 Fusion Protein Ag8510
Primary
-20°C
RPL29
0.14 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
60S ribosomal protein L29, HIP, HUMRPL29, ribosomal protein L29, RPL29
RPL29
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RPL29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.