missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RPS25 Recombinant Protein

Brand:  Abnova™ H00006230-P01.25ug

 View more versions of this product

Product Code. 16190382

  • 4040.00 DKK / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH03537
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
39.49
8
Glutathione Sepharose 4 Fast Flow
25 μg
MPPKDDKKKKDAGKSAKKDKDPVNKSGGKAKKKKWSKGKVRDKLNNLVLFDKATYDKLCKEVPNYKLITPAVVSERLKIRGSLARAALQELLSKGLIKLVSKHRAQVIYTRNTKGGDAPAAGEDA
RPS25
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
6230
RPS25 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPS25
Wheat Germ (in vitro)
GST
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers