missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(RS)-Baclofen, Tocris Bioscience™
Click to view available options
Quantity:
1 g
Description
Selective GABAB receptor agonist. Skeletal muscle relaxant.
R-enantiomer also available.
Specifications
Specifications
CAS | 1134-47-0 |
Molecular Formula | C10H12ClNO2 |
Synonym | baclofen, lioresal, baclon, 4-amino-3-4-chlorophenyl butanoic acid, dl-baclofen, kemstro, gabalon, baclofene, baclofeno, baclofenum |
InChI Key | KPYSYYIEGFHWSV-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1C(CC(=O)O)CN)Cl |
IUPAC Name | 4-amino-3-(4-chlorophenyl)butanoic acid |
Molecular Weight (g/mol) | 213.661 |
PubChem CID | 2284 |
ChEBI | CHEBI:2972 |
Formula Weight | 213.66 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'