missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPH9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-83479-0.1ML
This item is not returnable.
View return policy
Description
RSPH9 Polyclonal specifically detects RSPH9 in Human samples. It is validated for Western Blot.Specifications
RSPH9 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
221421 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
C6orf206, chromosome 6 open reading frame 206, CILD12, FLJ30845, mitochondrial ribosomal protein S18A-like 1, MRPS18AL1, radial spoke head 9 homolog (Chlamydomonas), radial spoke head protein 9 homolog | |
The immunogen is a synthetic peptide directed towards the middle region of Human RSPH9 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction