Learn More
Fisherbrand™ Cork Stoppers
For use in education, homebrewing and arts and crafts industries.
Brand:  Fisherbrand™ CB041.5/S/Z
 View more versions of this product
Product Code. 12396467
  • 119.00 DKK / Pack of 10
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Available in different sizes
Specifications
Specifications
32 mm
41.5 mm
Cork
10/Pk.
44 mm
Cork Stopper
Documents
One moment while we fetch your results.
Product Title
Fisherbrand™ Cork Stoppers > Base Diameter: 41.5mm