missing translation for 'onlineSavingsMsg'
Learn More
uvex Rubiflex NB60SZ Chemical Protection Gloves Product Code.: 17210411
Only undefined left
Add to basket

uvex Rubiflex NB60SZ Chemical Protection Gloves

Product Code. 17210411 Shop All Uvex Products
11
Click to view available options
Size:
9
10
11
This item is not returnable. View return policy

Product Code. 17210411

Brand: uvex 6018062

Only undefined left
Edge
Add to basket
Only undefined left
Add to basket
This item is not returnable. View return policy

Full NBR-coating reinforced with cotton interlock supporting material for applications that require protection against chemicals and mechanical risks. Elastic on the glove cuff provides additional protection by preventing hazards from entering the glove.

  • Extended length chemical protection glove with reinforced cotton interlock lining
  • Cotton liner helps absorb water vapor
  • Additional elastic collar at gauntlet end
  • Resistant to many chemicals, acids, alkalis, mineral oils, and solvents
  • Suitable for applications that require protection against chemicals and mechanical risks (ex. heat)
TRUSTED_SUSTAINABILITY

Specifications

Cuff Material Elastic
Applications Laboratory
Coating Material NBR
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 11
EN388 Standard 2121X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKOPT
For Use With (Application) Chemical Handling, Drainage Construction, and Municipal Cleaning
Gender Unisex
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Rubiflex NB60SZ
Length (Metric) 60 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
uvex Rubiflex NB60SZ Chemical Protection Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.