missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUVBL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10210-2-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody to human RUVBL1.
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It may attenuate protein kinase C activity by regulating diacylglycerol levels in intracellular signaling cascade and signal transduction. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified.Specifications
RUVBL1 | |
Polyclonal | |
Unconjugated | |
RUVBL1 | |
49 kDa TBP interacting protein, ECP 54, ECP54, INO80 complex subunit H, INO80H, Nuclear matrix protein 238, PONTIN, Pontin 52, Pontin52, RuvB like 1, RuvB like 1 (E. coli), RUVBL1, RVB1, TAP54 alpha, TIH1, TIP49, TIP49A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56505, 65137, 8607 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P60122, P60123, Q9Y265 | |
Ruvbl1 | |
RUVBL1 Fusion Protein Ag0281 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUVBL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction