Learn More
SCAT™ Safety Waste Caps
Safety Waste Caps for the safe disposal of liquid waste. For maximum protection and efficient work with hazardous waste liquids. Offers multiple options to connect different tubing and capillary sizes.
1045.00 DKK - 1905.00 DKK
Product Code | Brand | Size | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Includes | Price | Quantity & Availability | |||||
10681714
![]() |
SCAT™
107037 |
B53 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
1330.00 DKK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10529850
![]() |
SCAT™
107038 |
B63 | 1 x 2.3/3.2 mm OD, 3 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
1140.00 DKK
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
10880191
![]() |
SCAT™
107051 |
B63 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
10170510
![]() |
SCAT™
107050 |
B63 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
10785821
![]() |
SCAT™
107036 |
B83 | 4 x 2.3/3.2 mm OD Connector for Exhaust Filter |
1810.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
10619993
![]() |
SCAT™
107034 |
B83 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
1905.00 DKK
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
10119164
![]() |
SCAT™
108025 |
S50 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10614237
![]() |
SCAT™
108023 |
S50 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10377609
![]() |
SCAT™
108024 |
S50 | 3 x 2.3/3.2 mm OD, Grounding Connection, Connector for Exhaust Filter |
1525.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
10272723
![]() |
SCAT™
107922 |
S51 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10265103
![]() |
SCAT™
107930 |
S51 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10605645
![]() |
SCAT™
107924 |
S55 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
1045.00 DKK
Each |
In Stock
Log in to see stock. ![]() |
|||||
10699515
![]() |
SCAT™
107917 |
S55 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
10717595
![]() |
SCAT™
108047 |
S65 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
1330.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
10218784
![]() |
SCAT™
108046 |
S65 | 5 x 2.3/3.2 mm OD Connector for Exhaust Filter |
1330.00 DKK
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
10629993
![]() |
SCAT™
107947 |
S90 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
1330.00 DKK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
10508625
![]() |
SCAT™
107927 |
S90 | 4 x 2.3/3.2 mm OD Connector for Exhaust Filter |
1235.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
12655895
![]() |
SCAT™
107257 |
S95 | 4 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID Connector for Exhaust Filter |
1715.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
10580241
![]() |
SCAT™
107926 |
S70/71 | 2 x 2.3/3.2 mm OD, 1 x 5 to 11.5 mm ID, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
10861511
![]() |
SCAT™
107913 |
S70/71 | 3 x 2.3/3.2 mm OD, Connector for Exhaust Filter |
1045.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
17927250
|
SCAT™
108142 |
1415.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||||
17947260
|
SCAT™
108177 |
1320.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||||
Description
- 360° free turning PTFE body, resistant against aggressive media
- Exhaust filter system against harmful vapors
- Fittings and tube connectors in different sizes
- Flexible system
- Time saving in changing the waste containers while tubes are fixed
- Lab safety, technical protective measures, optimal protection when collecting liquid waste
- Cost efficiency
- Huge range of accessories, optional: funnel use, level control preventing overflowing containers
- Huge portfolio of thread sizes suitable for waste containers from different manufacturers
- Safety disposal of HPLC waste
- High work performance
- Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
- Sealed bottles and containers
- Create a comfortable working climate
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
SCAT™ Safety Waste Caps