missing translation for 'onlineSavingsMsg'
Learn More

S0859, MedChemExpress

MedChemExpress S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).

Encompass_Preferred

684.84 DKK - 4461.56 DKK

Specifications

Chemical Name or Material S0859
CAS 1019331-10-2
Color White
Purity Grade Notes Research
Molecular Formula C29H24ClN3O3S
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16486598
Encompass_Preferred
View Documents
Medchem Express
HY-15529
10 mg
1148.12 DKK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16469167
Encompass_Preferred
View Documents
Medchem Express
HY-15529
5 mg
684.84 DKK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16412697
Encompass_Preferred
View Documents
Medchem Express
HY-15529
50 mg
4461.56 DKK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

S0859 is a selective, high-affinity generic Na+/HCO3- transporter (NBC) inhibitor. S0859 reversibly inhibits NBC-mediated intracellular pH (pHi) recovery (Ki=1.7 μM, full inhibition at approximately 30 μM).
Specifications

Specifications

S0859
White
C29H24ClN3O3S
O=C(N(CC1=CC=C(C2=CC=CC=C2S(=O)(NC#N)=O)C=C1)CC3=CC=C(C)C=C3)C4=CC=CC=C4Cl
530.04
Research
COVID-19-immunoregulation
1019331-10-2
Research
DMSO : ≥ 100 mg/mL (188.67 mM)
530.04
98.0%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
S0859, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.