missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ S100A4 Recombinant Protein

Brand:  Abnova™ H00006275-P01.10ug

 View more versions of this product

Product Code. 16170432

  • 2665.00 DKK / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH16300
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
36.85
8
Glutathione Sepharose 4 Fast Flow
10 ug
MACPLEKALDVMVSTFHKYSGKEGDKFKLNKSELKELLTRELPSFLGKRTDEAAFQKLMSNLDSNRDNEVDFQEYCVFLSCIAMMCDEFFEGFPDKQPRKK
18A2/42A/CAPL/FSP1/MTS1/P9KA/PEL98
S100A4
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6275
S100A4 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
S100A4
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers