Learn More
Socorex™ Acura™ Electro 926 Micropipette
Lightweight and ergonomically friendly, this electronic single-channel micropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 926 Micropipette charges quickly and has a long working autonomy.
Brand: Socorex™ 926.0010
This item is not returnable.
View return policy
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
- Includes pipetting cycle counter.
- Optional volumetric modules.
Specifications
<±1.2% - <±0.6% | |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
1 | |
Self-teaching | |
0.5 to 10 μL (Ultra) | |
QC certificate, Qualitips™ pipette tip samples and operating instructions. | |
<1.5% - <0.35% | |
<1.5 Hours |
4.8V NiMH | |
926 | |
Acura Electro | |
Electronic Micropipette | |
Right/Left LCD | |
0.05μL | |
1 Ea. | |
24 Months |
Product Content Correction
Product Title
Socorex™ Acura™ Electro 926 Micropipette > Capacity: 0.5 to 10μL; Battery: 4.8V
Spot an opportunity for improvement?Share a Content Correction