missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Glucose Chloramphenicol Selective Agar Contact Plate

Isolate, cultivate and differentiate dermatophytes, fungi and yeats using this formulation in contact plate format.

Brand:  Thermo Scientific™ PO5094C

162.59 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10690005

  • 179.00 DKK / Pack of 10
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Honey yellow, transparent
Sabouraud Glucose Chloramphenicol Selective Agar Contact Plate
Germany
13.5g ± 5 %
2 x 10/Pk.
2 to 25°C
Contact Plate
Agar
Boxes with 2 x 10 plates wrapped in film
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6276373 Certificate TypeCertificate of Analysis Date03/06/2025 Catalog Number
Lot Number6273796 Certificate TypeCertificate of Analysis Date26/05/2025 Catalog NumberPO5094C
Lot Number6270859 Certificate TypeCertificate of Analysis Date20/05/2025 Catalog NumberPO5094C
Lot Number6268233 Certificate TypeCertificate of Analysis Date14/05/2025 Catalog NumberPO5094C
Lot Number6260956 Certificate TypeCertificate of Analysis Date23/04/2025 Catalog NumberPO5094C
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Sabouraud Glucose Chloramphenicol Selective Agar Contact Plate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.