Learn More
Mettler Toledo™ Sachet Holder, EasyPlace, Rotating
The sachet holder can hold up to three buffer sachets and prevent spillage. It can be rotated to place one buffer after the other in the calibration position.
Brand: Mettler Toledo™ 30671573
285.38 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Safe Sachet Handling
- The rotating sachet holder sits securely on the EasyPlace baseplate. It holds up to three standard sachets, preventing spills
- Straightforward Calibration
- The placement on the EasyPlace makes sensor insertion simple. A simple turn of the holder moves the next standard into the measurement position
Specifications
Sachet Holder, EasyPlace Rotating | |
EasyPlace |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.