Learn More
Brady™ Safety Sign without Text - No Access for Unauthorized Persons
Safety sign for indicating mandatory action.
57.90 DKK - 118.00 DKK
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | ||||||
15604207
|
Brady™
222923 |
100 mm |
57.90 DKK
Each |
In Stock
Log in to see stock. |
||||||
16200051
|
Brady™
255016 |
200 mm |
118.00 DKK
Each |
Estimated Shipment: 11-07-2025
Log in to see stock. |
||||||
Description
- Improves workplace safety
Specifications
Round | |
Red on White |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brady™ Safety Sign without Text - No Access for Unauthorized Persons