Learn More
SymLine™ Safety Waste Caps
These Safety Waste Caps safely connect the pipework to the waste container as an integrated exhaust filter blocks solvent vapors for health and environmental protection.
Brand:  SymLine™ 106720
4157.18 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17838743
  • 5280.00 DKK / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Description
  • Available with different thread sizes for attachment to pipe system
  • Different numbers of hose connections for disposal of liquid waste
  • Available with electronic level sensors that indicate the degree of filling of the disposal container
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
Specifications
Specifications
Black
SymLine System
PTFE-EC
Safety Waste Cap
Waste Disposal
Electronic Level Control with Shut-off GL25 (m), Connector for Exhaust Filter
S90