Learn More
Salidroside, MedChemExpress
MedChemExpress Salidroside is a prolyl endopeptidase inhibitor. Salidroside alleviates cachexia symptoms in mouse models of cancer cachexia via activating mTOR signalling. Salidroside protects dopaminergic neurons by enhancing PINK1/Parkin-mediated mitophagy.

684.84 DKK - 1107.84 DKK
Specifications
Chemical Name or Material | Salidroside |
---|---|
CAS | 10338-51-9 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C14H20O7 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16448868
![]() |
Medchem Express
HY-N0109 |
10 mg |
1107.84 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431308
![]() |
Medchem Express
HY-N0109 |
5 mg |
684.84 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Salidroside is a prolyl endopeptidase inhibitor. Salidroside alleviates cachexia symptoms in mouse models of cancer cachexia via activating mTOR signalling. Salidroside protects dopaminergic neurons by enhancing PINK1/Parkin-mediated mitophagy.Specifications
Salidroside | |
Off-White | |
C14H20O7 | |
H2O : ≥ 100 mg/mL (333.00 mM) | |
300.3 | |
98.0% | |
Solid |
10338-51-9 | |
Research | |
Rhodioloside | |
O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1OCCC2=CC=C(O)C=C2 | |
300.3 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
H319
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.