Learn More
Hamilton™ SaltLine™ Luer Lock Syringes
Choose these syringes when a precise flow is critical for almost any application.
Brand:  Hamilton™ 203240
 View more versions of this product
Product Code. 11522495
  • 1535.00 DKK / Each
Explore more special offers
Description
Description
  • PTFE luer lock termination is extremely common as it provides the most flexible options for connection of tubing
  • Available in a range of volumes
  • Applications include microfluidics, drug/nutritional infusions, electrospinning, mass spectrometer calibration, cell feeding and more
Specifications
Specifications
Instrument Syringe
1/Ea.
Product Title
Hamilton™ SaltLine™ Luer Lock Syringes > Volume: 1mL