Learn More
Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes
Perform precise dispensing of volumes from 20 to 400 ÎĽL by using these exact volume transfer pipettes. Made of shatterproof, nontoxic polyethylene.
Brand: Thermo Scientific™ 786NL
2001.45 DKK valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
Perform precise dispensing of volumes from 20 to 400 μL by using Thermo Scientific™ Samco™ Exact Volume Transfer Pipettes. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
- Shatterproof, nontoxic polyethylene
- Nonsterile
- Low affinity surface
- Freezer ready
Specifications
80ÎĽL Exact Volume Transfer Pipette | |
LDPE | |
No | |
80 ÎĽL | |
Exact Volume Transfer Pipet |
2.5 mm | |
Non-sterile | |
Bulk 500/Pk 10Pk/Cs 5000/Cs | |
7.5 cm | |
Single-use |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Thermo Scientific Samco Exact volume tranfer pipettes
-
Thermo Scientific Samco Exact volume tranfer pipettes
Special Offers
-
Multiple Manufacturers
Grow Your Discovery
Fall in love with big savingsPromo Code: 24107Expires: 2025-03-28
Days
10Hours
18Minutes
34Seconds
11