Learn More
S Murray and Co™ SAMCO™ Household Scissors
Use these scissors for cutting anything from bandages to stainless steel mesh. S Murray™ SAMCO™ Household Scissors are heavy duty but lightweight with stainless steel blades and robust construction.
56.50 DKK - 64.55 DKK
Specifications
Product Type | Household Scissors |
---|---|
Blade Style | Straight |
Material | Stainless Steel |
Autoclavable | Autoclavable |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
12932065
|
S Murray and Co™
E901/04 |
150 mm |
56.50 DKK
Each |
Estimated Shipment: 18-08-2025
Log in to see stock. |
||||||
12942065
|
S Murray and Co™
E901/06 |
175 mm |
64.55 DKK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
||||||
Description
- General purpose, nickel plated steel household scissors
- Straight with one sharp point
- Autoclavable
Specifications
Household Scissors | |
Straight | |
Stainless Steel | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
S Murray and Co™ SAMCO™ Household Scissors