Learn More
S Murray and Co™ SAMCO™ Stainless Steel Scissors
Use these scissors for a wide range of applications. S Murray™ SAMCO™ Stainless Steel Scissors are available in a variety of blade and tip styles to meet your needs.
44.30 DKK - 76.40 DKK
Product Code | Brand | Length (Metric) | Blade Style | Tip Style | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Blade Style | Tip Style | Price | Quantity & Availability | ||||
12358099
|
S Murray and Co™
E802/02 |
125 mm | Straight | 1 Pointed, 1 Blunt |
44.30 DKK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||
12348099
|
S Murray and Co™
E828/01 |
115 mm | Straight | Very Fine Point |
76.40 DKK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||
12912055
|
S Murray and Co™
E803/02 |
125 mm | Straight | Fine Point |
50.35 DKK
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
Hai giĂ un account? Accedi per visualizzare il prezzo e la disponibilitĂ . Sei nuovo da queste parti? Registrati qui! |
|||
12494070
|
S Murray and Co™
E803/04 |
150 mm | Straight | Fine Point |
59.75 DKK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
Hai giĂ un account? Accedi per visualizzare il prezzo e la disponibilitĂ . Sei nuovo da queste parti? Registrati qui! |
|||
Descrizione
- Fully guaranteed in normal use and subject to stringent quality inspection procedures
Specifica
Stainless Steel Scissors | |
Stainless Steel |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.