Learn More
Kartell™ Sample Tube
For sample preparation, centrifugation, and for Autosampler racks
Brand:  Kartell™ 0167100
Product Code. 10316111
  • 1010.00 DKK / Each
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Description
  • Calibrated ring-mark at 10mL
  • Screw cap (PFA) GL 25
  • Material: PFA
  • Specifications
    Specifications
    PFA
    110 mm
    22 mm
    15 mL
    Plastic
    Screw Cap
    Documents
    One moment while we fetch your results.
    Product Title
    Kartell™ Sample Tube > Capacity (Metric): 15mL