Learn More
SARS-CoV-2 nsp9 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-07044AF532
Product Code. 18310942
  • 4848.69 DKK / 0.10mL
Explore more special offers
This item is not returnable. View return policy
Description
Description
SARS-CoV-2 nsp9 Polyclonal antibody specifically detects SARS-CoV-2 nsp9 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
SARS-CoV-2 nsp9
Polyclonal
50mM Sodium Borate with 0.05% Sodium Azide
Rabbit
Peptide Affinity-purified
RUO
Primary
SARS-CoV-2
Purified
ELISA, Immunohistochemistry, Immunofluorescence
Alexa Fluor 532
Non-structural protein 9, nsp9 replicase
This antibody was raised against a peptide corresponding to 13 amino acids near the center of SARS-CoV-2 (COVID-19) NSP9 protein. The immunogen is located within 30-80 amino acids of the SARS-CoV-2 (COVID-19) NSP9 protein.
0.1 mL
Infections (Virus Bacteria and Parasites)
43740578
Store at 4°C in the dark.
IgG