Learn More
Sartorius Balance Density Determination Kits
Use with Sartorius balances to determine the density of solids and liquids
Brand: Sartorius YDK01LP
8729.32 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
All kits include
- 2 Beakers (7mm and 55mm dia.)
- Bar frame
- Thermometer with retainer clip
- Glass plummet
- Sieve for immersing samples (pan hanger assembly)
- Sample holder (pan hanger assembly)
- Metal platform
- Gasket for ME models
- Compensating disk for ME235S/P, ME254S models
- 3 adapters
- Adapter for LA/LP models with analytical draft shield
- Adapters for LA/LP models: 1 with and 1 without analytical draft shield
- Compensating disk for LA/LP2300D models
Specifications
Extend ED, LA, LP Precision Balance | |
Determining the density of solids and liquids | |
2 Beakers (7 mm and 55 mm dia.), Bar Frame, Thermome | |
Density Determination Kit |
Product Content Correction
Product Title
Sartorius Balance Density Determination Kits > Extend, ED 224S
Spot an opportunity for improvement?Share a Content Correction