Learn More
Sartorius Remote Display for Sartorius Balances
For use with CPA Balances, Extend ED Balances, M-Power Balances.
Brand: Sartorius YRD03Z
2730.71 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Connect to balances using RS232 cable
- LCD displays come in reflective, transmissive or backlit models
Specifications
CPA Balance, Extended Balance, M-power Balance | |
Backlit LCD | |
Remote Display |