Learn More
Sartorius Replacement Part for Polycarbonate Filtration Holders
Replacement Part for Polycarbonate Filtration Holders is for use with Microsart™ Manifolds 16510 and 16511.
Brand: Sartorius 6980230
154.44 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polycarbonate Accessory
- For use with Microsart™ Manifolds 16510, 16511
Specifications
Reusable | |
Polycarbonate | |
Non-sterile | |
Top Part |
Filter Holder | |
PES | |
1 Ea. | |
250 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Sartorius Replacement Part for Polycarbonate Filtration Holders > Replacement Part for Polycarbonate Filtration Holders; Type: Top Part; Quantity: 1 Ea.
Spot an opportunity for improvement?Share a Content Correction