Learn More
Sartorius Vivaspin™ 500 Centrifugal Concentrators
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
849.52 DKK - 5125.00 DKK
Specifications
Disposable | Single-use |
---|---|
Filter Type | Ultrafiltration |
For Use With (Equipment) | Centrifuge |
Material (Membrane) | PES |
Sample Volume | 500 mL |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | |||||
10019460
![]() |
Sartorius
VS0101 |
10 kDa | 25/Pk. |
1250.00 DKK
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10719001
![]() |
Sartorius
VS0112 |
5 kDa | 100/Pk. |
4260.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10029510
![]() |
Sartorius
VS0111 |
5 kDa | 25/Pk. |
1280.00 DKK
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10125580
![]() |
Sartorius
VS0102 |
10 kDa | 100/Pk. |
4225.00 DKK
Pack of 100 |
In Stock
Log in to see stock. ![]() |
|||||
10681842
![]() |
Sartorius
VS0121 |
30 kDa | 25/Pk. |
1250.00 DKK
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10029470
![]() |
Sartorius
VS0131 |
50 kDa | 25/Pk. |
1335.00 DKK
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10375361
![]() |
Sartorius
VS0141 |
100 kDa | 25/Pk. |
1250.00 DKK
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10459512
![]() |
Sartorius
VS0132 |
50 kDa | 100/Pk. |
5125.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10073930
![]() |
Sartorius
VS0122 |
30 kDa | 100/Pk. |
5125.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10499701
![]() |
Sartorius
VS0151 |
300 kDa | 25/Pk. |
1465.00 DKK
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10223621
![]() |
Sartorius
VS0142 |
100 kDa | 100/Pk. |
4260.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10767052
![]() |
Sartorius
VS0152 |
300 kDa | 100/Pk. |
4170.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10727632
|
Sartorius
VS0161 |
1000 kDa | 25/Pk. | N/A | ||||||
10174281
|
Sartorius
VS0162 |
1000 kDa | 100/Pk. |
3430.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10184281
![]() |
Sartorius
VS0171 |
0.2 ÎĽm | 25/Pk. |
1560.00 DKK
Pack of 25 |
Estimated Shipment: 07-08-2025
Log in to see stock. |
|||||
10658961
![]() |
Sartorius
VS0172 |
0.2 ÎĽm | 100/Pk. |
4260.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10440282
![]() |
Sartorius
VS0191 |
3 kDa | 25/Pk. |
1280.00 DKK
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10488742
|
Sartorius
VS01S1 |
5, 10, 30, 50, and 100 kDa | 25/Pk. | N/A | ||||||
10689151
![]() |
Sartorius
VS0192 |
3 kDa | 100/Pk. |
5125.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
- Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
- Concentrate recoveries of > 90% can be reached in less than 30 minutes
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
- High-flux PES membranes for ultra-fast concentration
- Convenient concentrate recovery by pipetting
- Clear housing with volume graduation for easy reading/monitoring
Specifications
Single-use | |
Centrifuge | |
500 mL | |
64.5m m2 | |
50 mm |
Ultrafiltration | |
PES | |
Non-sterile | |
Polycarbonate |