Learn More
Tocris Bioscience™ SB 205607 dihydrobromide
Selective, high affinity non-peptide δ1 agonist
Brand: Tocris Bioscience™ 0921/10
2046.01 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The first described non-peptide δ1 opioid receptor agonist with very high affinity and selectivity for the δ1 subtype (Ki values are 1.12, 2320 and 1790 nM at δ1, μ and κ receptors respectively).Specifications
>97% | |
10 mg | |
506.28 |
TAN 67 | |
SB 205607 dihydrobromide | |
SB 205607 dihydrobromide |
For Research Use Only