Learn More
DWK Life Sciences Screw Cap, for Culture tubes, with seal
This screw cap is used to close culture tubes with threads (threads not according to DIN).
Brand: DWK Life Sciences 299901307
136.82 DKK valid until 2024-10-31
Use promo code "21661" to get your promotional price.
Description
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today's research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. In addition to culture tubes made of DURAN™, our product range also includes culture tubes made of soda-lime glass. This is a glass of the third water resistance Class And belongs to the category of soda-lime glasses with a high proportion of alkali and alkaline earth oxides.
- Cultivating, storage and transport of microorganisms
Specifications
GL18 | |
Polypropylene, TPE | |
100/Pk. |
Autoclavable | |
Growth of microorganisms; Storage and transport of small quantities of liquid; Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods |