Learn More
BD Select APS Tryptic Soy Broth
BD Select APS Tryptic Soy Broth is used in microbiology laboratories as a culture broth to grow bacteria.
Brand: BD 214886
This item is not returnable.
View return policy
Description
- Tryptic soy broth is used in microbiology laboratories as a culture broth to grow aerobic bacteria.
- Used in a wide variety of procedures
- May be used for suspension and enrichment
- For cultivation of strains isolated on other media
- With the addition of 6.5% sodium chloride, differentiation between salt-tolerant and salt-intolerant species is achieved.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
TSB | |
Blue | |
10 kg |