Learn More
Thermo Scientific™ Sensors and Replacement Batteries for Thermo Scientific™ Ultra-Low Temperature Freezers
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
Brand: Thermo Scientific™ AVL400159
619.20 DKK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
Specifications
Replacement Back-Up Battery | |
Forma™900, Forma™ 8600 Series, Forma™ 88000, TSD Series, TSUSeries |
Product Suggestions
Customers who viewed this item also viewed.
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
73Hours
19Minutes
41Seconds
48New Offer Save Up to 59% -
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
New Offer Save Now -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
18Hours
19Minutes
41Seconds
48