Learn More
Fisherbrand™ Septum for 13-425 screw cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 12 02 0444
Description
- 12mm Septa
Specifications
Septa | |
12 mm | |
1000 |
Red-Orange, Transparent | |
Natural Rubber, TEF | |
1 mm |