missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertraline hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, orally active selective serotonin re-uptake inhibitor (SSRI); antidepressant.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 342.69 |
Percent Purity | >99% |
Chemical Name or Material | Sertraline hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'