Learn More
Vacuubrand™ Set Of Seals
Brand: Vacuubrand™ 20639123
139.65 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
For Oil Separator FO R2, R6 | |
1 Each | |
Seal Set |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
12Hours
20Minutes
59Seconds
50New Offer Save Up to 92%
Product Content Correction
Product Title
Vacuubrand™ Set Of Seals > For use with: For Oil Separator FO R2, R6
Spot an opportunity for improvement?Share a Content Correction