missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sex Hormone Binding Globulin Mouse, Clone: SHBG/245, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6462-MSM1-P1
This item is not returnable.
View return policy
Specifications
Sex Hormone Binding Globulin | |
Cytoplasmic and Extracellular (Secreted) | |
SHBG/245 | |
Unconjugated | |
NP_001276044 | |
Mouse | |
45kDa | |
Protein A/G Purified | |
RUO | |
6462 | |
IgG1 |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide | |
SHBG | |
Recombinant full-length human SHBG protein | |
Liver, Placenta or Testis | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sex Hormone Binding Globulin Mouse, Clone: SHBG/245, enQuireBio™ > 100 μg
Spot an opportunity for improvement?Share a Content Correction