missing translation for 'onlineSavingsMsg'
Learn More

SHA 68, Tocris Bioscience™

Product Code. 30001677
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective neuropeptide S receptor antagonist

Selective neuropeptide S receptor (NPSR) antagonist (IC50 values are 22.0 and 23.8 nM for human NPSR Asn107 and Ile107 variants respectively). Displays no activity against a range of 14 GPCRs, including vasopressin and oxytocin receptors.

Chemical Identifiers

CAS 847553-89-3
Molecular Formula C26H24FN3O3
Molecular Weight (g/mol) 445.494
InChI Key SFRQIPRTNYHJHP-UHFFFAOYSA-N
Synonym sha 68, sha-68, sha68, n-4-fluorobenzyl-3-oxo-1,1-diphenyltetrahydro-1h-oxazolo 3,4-a pyrazine-7 3h-carboxamide, n-4-fluorophenyl methyl tetrahydro-3-oxo-1,1-diphenyl-3h-oxazolo 3,4-a pyrazine-7 1h-carboxamide, d03ejp, n-4-fluorophenyl methyl-3-oxo-1,1-di phenyl-5,6,8,8a-tetrahydro-1,3 oxazolo 4,3-c pyrazine-7-carboxamide, n-4-fluorophenyl methyl-3-oxo-1,1-diphenyl-tetrahydro-1,3 oxazolo 3,4-a pyrazine-7-carboxamide
PubChem CID 11374217
IUPAC Name N-[(4-fluorophenyl)methyl]-3-oxo-1,1-diphenyl-5,6,8,8a-tetrahydro-[1,3]oxazolo[3,4-a]pyrazine-7-carboxamide
SMILES C1CN2C(CN1C(=O)NCC3=CC=C(C=C3)F)C(OC2=O)(C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Quantity 50 mg
Formula Weight 445.49
Percent Purity >98%
Chemical Name or Material SHA 68
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.