Learn More
Memmert™ Shaking Device for Waterbaths
Shaking device with support frame without gable cover & support basket (mandatory)
11420.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17110415
|
Memmert™
WTB50SV3550 |
WTB50 |
11420.00 DKK
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
|||||||||
17180385
|
Memmert™
WTB35SV3550 |
WTB35 |
11420.00 DKK
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
- Requires gable cover
- Voltage 230 V, 50/60 Hz
- Order the support basket separately
- Includes support frame, shaking frequency 35 - 160 strokes per minute (horizontal back/forth movement)
Specifications
Shaking Device | |
35-160 rpm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Memmert™ Shaking Device for Waterbaths