Learn More
Epredia™ Cryochrome™ Embedding Resin
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
675.00 DKK - 841.00 DKK
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12533527
|
Epredia™
9990422 |
Cryochrome Blue | |||||||
12560017
|
Epredia™
9990423 |
Cryochrome Yellow | |||||||
12726087
|
Epredia™
9990426 |
Cryochrome Sampler Colors | |||||||
12570017
|
Epredia™
9990426 |
Cryochrome Sampler Colors |
841.00 DKK
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12543527
|
Epredia™
9990428 |
Cryochrome Green | |||||||
12580017
|
Epredia™
9990429 |
Cryochrome Pink | |||||||
Description
- Color codes frozen sections for easy identification and tracking
- Four colors available: pink, blue, yellow, green
- Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
120mL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Epredia™ Cryochrome™ Embedding Resin