Learn More
Epredia™ Shandon™ Gill™ Hematoxylin
Achieve fast and convenient results, while maintaining complete control over stain intensity with Epredia™ Shandon™ Gill™ Hematoxylin, which has mucoid staining.
Brand: Epredia™ 6765010
991.20 DKK valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Gill 1: Light blue nuclei
Gill 2: Medium blue nuclei
Gill 3: Blue-black nuclei
Specifications
Shandon Gill™ 3 Hematoxylin | |
4L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- SHANDON GILL 3 HAEMATOXYLIN CE-IVD 4L .
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Ethylene glycol, Acetic acid