Learn More
Epredia™ Glyo-Fixx™
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
Brand:  Epredia™ 6764260
 View more versions of this product
Product Code. 10249908
  • 1750.00 DKK / Each
Explore more special offers
Description
Description
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fixative
5 gal.
Documents
One moment while we fetch your results.
Product Title
Epredia™ Glyo-Fixx™ > 5 gal. (18.9L); 1 ea.