Learn More
Epredia™ Shandon™ Polysine Slides, White Product Code.: 12618106
Epredia™ Shandon™ Polysine Slides, White
Product Code. 12618106 Shop All Epredia Products
White; 1440/Case, Pack of 1440
Click to view available options
Description:
Blue; 144/Pack
White; 144/Pack
White; 1440/Case
Unit Size:
Pack of 144
Pack of 1440
Product Code. 12618106
Brand: Epredia™ 6776215CS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve superior cell and tissue adhesion with Epredia™ Shandon™ Polysine Slides.
  • Polysine places an advanced permanent adhesive on the microscope slide
  • Built-in chemicals electrostatically and chemically attract tissues and cells
TRUSTED_SUSTAINABILITY
Specifications
Material Polysine
Type Microscope Slide
Color White
Description White; 1440/Case
Product Content Correction
Product Title
Epredia™ Shandon™ Polysine Slides, White >